1,2-Dibromoethane 99%

Danger

AC11279-0025 AC11279-0010 AC11279-0100
103524-878EA 280 USD
103524-878 103524-876 103524-880
1,2-Dibromoethane 99%
1,2-Dibromoethane
Formula: BrH₂CCH₂Br
MW: 187.86 g/mol
Boiling Pt: 131…132 °C (760 mmHg)
Melting Pt: 9…10 °C
Density: 2.173 g/cm³
Flash Pt: >104 °C
Storage Temperature: Ambient
MDL Number: MFCD00000233
CAS Number: 106-93-4
EINECS: 203-444-5
UN: 1605
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5375 to 1.5395 (20 °C, 589 nm)
GC ≥98.5 %
Specific optical rotation -29° to -33° (20 °C, 589 nm) (c=1, toluene)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR