1,2-Bis(dicyclohexylphosphino)ethane 98%

Warning

AC36385-0050
103512-736EA 315 USD
103512-736
1,2-Bis(dicyclohexylphosphino)ethane 98%
1,2-Bis(dicyclohexylphosphino)ethane
Formula: C₂₆H₄₈P₂
Storage Temperature: Ambient
MDL Number: MFCD00015521
CAS Number: 23743-26-2

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Assay ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR