1,1-Diphenyl-2-propyn-1-ol 98%

Synonyms: 1,1-Diphenylprop-2-yn-1-ol

AC38997-0250 AC38997-0050
103513-114EA 39.7 USD
103513-114 103513-116
1,1-Diphenyl-2-propyn-1-ol 98%
1,1-Diphenyl-2-propyn-1-ol
Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Boiling Pt: 183 °C (20 mmHg)
Melting Pt: 46…50 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00041570
CAS Number: 3923-52-2

Order Now

Specification Test Results

Appearance White to cream to light yellow Powder or crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 44 to 50 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR