1,1'-Bis(di-tert-butylphosphino)ferrocene 98%

Warning

Synonyms: 1,1'-Bis(di-t-butylphosphino)ferrocene1,1'-Bis[bis(1,1-dimethylethyl)phosphino]ferroceneDTBPF

AC42971-0010
103515-112EA 118.46 USD
103515-112
1,1'-Bis(di-tert-butylphosphino)ferrocene 98%
1,1'-Bis(di-tert-butylphosphino)ferrocene
Formula: C₂₆H₄₄FeP₂
MW: 474.42 g/mol
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD01630818
CAS Number: 84680-95-5

Order Now

Specification Test Results

Appearance Orange to red Powder
Infrared spectrum Conforms
Melting point 73 to 76 °C
NMR (31P-NMR) consistent with structure
Elemental Analysis 64.18 to 67.47 % C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR