EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 97%

Danger

Synonyms: EDACN'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamineEDCWSC1-(3-Dimethylaminopropyl)-3-ethylcarbodiimideN1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine

AC43884-0500 AC43884-0100
103524-464EA 715 USD
103524-464 103524-462
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 97%
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide
Formula: C₈H₁₇N₃
MW: 155.24 g/mol
Boiling Pt: 66…68 °C (1 mmHg)
Density: 0.885 g/cm³
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00044916
CAS Number: 1892-57-5
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4630 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR