(S)-N-FMOC-α-Methylphenylalanine sesquihydrate 98%, 98% ee

Synonyms: Fmoc-α-methyl-L-phenylalanine 3/2H2O

AC44108-1000
103516-124EA 241 USD
103516-124
(S)-N-FMOC-α-Methylphenylalanine sesquihydrate 98%, 98% ee
(S)-N-FMOC-alpha-Methylphenylalanine sesquihydrate
Formula: C₂₅H₂₃NO₄₁·5H₂O
MW: 428.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015046
CAS Number: 135944-05-7

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Enantiomeric excess ≥98.0 %
Specific optical rotation -28° to -32° (20 °C, 589 nm) (c=1 in DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR