(S)-N-Boc-allylglycine 95%

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)pent-4-enoic acid(S)-2-Boc-AMino-4-pentenoic acid4-Pentenoic acid, 2-[[(1,1-dimethylethoxy)carbonyl]amino]-, (2S)-

AC39010-5000
103513-970EA 123 USD
103513-970
(S)-N-Boc-allylglycine 95%
(S)-N-Boc-allylglycine
Formula: C₁₀H₁₇NO₄
MW: 215.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320851
CAS Number: 90600-20-7

Order Now

Specification Test Results

Appearance Colorless to yellow Viscous liquid or paste
Infrared spectrum Conforms
Specific optical rotation +9.0° to +13.0° (20 °C, 589 nm) (c=1, methanol)
Titration with NaOH ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR