L-Propargylglycine 98%, 98% ee

Synonyms: (S)-alpha-Propargylglycine(S)-2-Aminopent-4-ynoic acidH-Pra-OH

AC44121-5000 AC44121-1000
103513-320EA 802 USD
103513-320 103516-144
L-Propargylglycine 98%, 98% ee
L-Propargylglycine
Formula: C₅H₇NO₂
MW: 113.116 g/mol
Melting Pt: >180 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00077855
CAS Number: 23235-01-0

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Assay 97.5 to 102.5 %
Enantiomeric excess ≥98.0 %
Specific optical rotation -31° to -35° (20 °C, 589 nm) (c=1 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR