2-Methylleucine 98%, 98% ee

Synonyms: H-α-Me-Leu-OH(S)-2-Amino-2,4-dimethylpentanoic acid(S)-alpha-Methylleucine

AC44126-1000
103513-436EA 158 USD
103513-436
2-Methylleucine 98%, 98% ee
2-Methylleucine
Formula: C₇H₁₅NO₂
MW: 145.202 g/mol
Melting Pt: >237 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01716279
CAS Number: 105743-53-1

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Assay 97.5 to 102.5 %
Enantiomeric excess ≥98.0 %
Specific optical rotation +33° to +39° (20 °C, 589 nm) (c=1 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR