(S)-1-Boc-3-hydroxypiperidine 97%

Warning

Synonyms: (S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate(3S)-3-Hydroxypiperidine, N-BOC protected

AC43644-0010
103515-810EA 89.3 USD
103515-810
(S)-1-Boc-3-hydroxypiperidine 97%
(S)-1-Boc-3-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 48.9…51.7 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD04115307
CAS Number: 143900-44-1

Order Now

Specification Test Results

Appearance White to tan Powder
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation +9° to +11° (20 °C, 589 nm) (C=1 in chloroform) / +21° to +24° (20 °C, 589 nm) (c=5 in MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR