(S)-(+)-3-Aminopiperidine dihydrochloride 97%

Warning

Synonyms: (S)-(+)-3-Piperidinamine dihydrochloride(S)-Piperidin-3-amine dihydrochloride

AC43652-0050 AC43652-0010
103515-828EA 352.7 USD
103515-828 103515-826
(S)-(+)-3-Aminopiperidine dihydrochloride 97%
(S)-(+)-3-Aminopiperidine dihydrochloride
Formula: C₅H₁₄Cl₂N₂
MW: 173.08 g/mol
Melting Pt: 217.2…217.9 °C
Storage Temperature: Ambient
MDL Number: MFCD03427036
CAS Number: 334618-07-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Additional info Optical purity > 97%
Elemental Analysis 33.31 to 36.09 % C
Elemental Analysis 15.53 to 16.83 % N
Specific optical rotation +15° to +25° (20 °C, 589 nm) (c=1, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR