(S)-4-Hydroxypyrrolidine-2-one 98%

Warning

Synonyms: (S)-4-Hydroxy-2-pyrrolidone(S)-(-)-4-Hydroxy-2-pyrrolidinone(4S)-(-)-beta-Hydroxy-gamma-butyrolactam

AC39897-0010
103514-540EA 102 USD
103514-540
(S)-4-Hydroxypyrrolidine-2-one 98%
(S)-4-Hydroxypyrrolidine-2-one
Formula: C₄H₇NO₂
MW: 101.105 g/mol
Melting Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00273370
CAS Number: 68108-18-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 156 to 159 °C
GC ≥97.5 %
Specific optical rotation -40° to -46° (20 °C, 589 nm) (c=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR