(S)-(-)-2-(Boc-amino)-1,4-butanediol 97%

Warning

Synonyms: (S)-N-Boc-2-Aminobutane-1,4-dioltert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate(S)-2-(tert-butoxycarbonylamino)-1,4-butanediol

AC37510-0010 AC37510-0050
103512-766EA 267 USD
103512-766 103512-764
(S)-(-)-2-(Boc-amino)-1,4-butanediol 97%
(S)-(-)-2-(Boc-amino)-1,4-butanediol
Formula: C₉H₁₉NO₄
MW: 205.254 g/mol
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD03093946
CAS Number: 128427-10-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 65 to 69 °C
GC ≥96.0 %
Enantiomeric excess ≥98.0 %
Specific optical rotation -7° to -9° (20 °C, 589 nm) (c=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR