(R)-N-Fmoc-α-methylphenylalanine sesquihydrate 98%, 98% ee

Synonyms: (R)-N-FMOC-α-Methylphenylalanine sesquihydrate(R)-N-FMOC-α-METHYLPHENYLALANINE 3/2H2O

AC44107-1000
103516-122EA 240.9 USD
103516-122
(R)-N-Fmoc-α-methylphenylalanine sesquihydrate 98%, 98% ee
(R)-N-Fmoc-alpha-methylphenylalanine sesquihydrate
Formula: C₂₅H₂₃NO₄1.5H₂O
MW: 428.48 g/mol
Melting Pt: 79…104 °C
Storage Temperature: Ambient
MDL Number: MFCD17019321
CAS Number: 152436-04-9

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Enantiomeric excess ≥98.0 %
Specific optical rotation +28° to +32° (20 °C, 589 nm) (c=1 in DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR