(R)-α-Methylaspartic acid 98%, 98% ee

Synonyms: (2R)-2-amino-2-methylbutanedioic acid(R)-(-)-2-AMINO-2-METHYLBUTANEDIOIC ACIDD-Aspartic acid,2-methyl-, hydrochloride (1:1)

AC29014-5000 AC29014-1000
103512-060EA 350.6 USD
103512-060 103512-058
(R)-α-Methylaspartic acid 98%, 98% ee
(R)-alpha-Methylaspartic acid
Formula: C₅H₉NO₄
MW: 147.13 g/mol
Melting Pt: >249 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00216906
CAS Number: 14603-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR