(R)-3-Phenylmorpholine hydrochloride 97%

Warning

AC45679-0050 AC45679-0010
103518-420EA 191.4 USD
103518-420 103518-422
(R)-3-Phenylmorpholine hydrochloride 97%
(R)-3-Phenylmorpholine hydrochloride
Formula: C₁₀H₁₃NO·ClH
MW: 199.68 g/mol
Melting Pt: 192…193.5 °C
Storage Temperature: Ambient
MDL Number: MFCD12910599
CAS Number: 1624261-27-3

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 to ≤104.0 %
HPLC ≥96.0 %
Specific optical rotation -1.0° to -3.0° (20 °C, 589 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR