(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride monohydrate 97%

Warning

Synonyms: D-(+)-Homoserine Lactone Hydrochloride(R)-(+)-3-Aminotetrahydrofuran-2-one Hydrochloride

AC43757-0010 AC43757-0050
103515-952EA 345.2 USD
103515-952 103515-950
(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride monohydrate 97%
(R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride monohydrate
Formula: C₄H₇NO₂·HCl
MW: 137.56 g/mol
Melting Pt: 215.1…217.7 °C
Storage Temperature: Ambient
MDL Number: MFCD26793416
CAS Number: 104347-13-9

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 to ≤104.0 %
Water ≤15 %
Specific optical rotation +25° to +30° (20 °C, 589 nm) (c=1 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR