(R)-1-Benzyl-3-pyrrolidinol 98%

Warning

Synonyms: (R)-1-Benzyl-3-hydroxypyrrolidine(R)-3-Hydroxy-1-benzylpyrrolidine(R)-(+)-1-Benzyl-3-pyrrolidinol

AC44685-0050 AC44685-0010
103516-678EA 192.2 USD
103516-678 103516-676
(R)-1-Benzyl-3-pyrrolidinol 98%
(R)-1-Benzyl-3-pyrrolidinol
Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Boiling Pt: 88…90 °C (1Torr)
Storage Temperature: Refrigerator
MDL Number: MFCD00075484
CAS Number: 101930-07-8

Order Now

Specification Test Results

Appearance Colorless to yellow to brown Viscous liquid
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation +2.8° to +4.8° (20 °C, 589 nm) (c=5 in MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR