(R)-(-)-3-Aminopiperidine dihydrochloride 97%

Warning

Synonyms: (R)-(-)-3-Piperidinamine dihydrochloride(R)-Piperidin-3-amine dihydrochloride

AC43641-2500 AC43641-0010
103515-804EA 34.2 USD
103515-804 103515-802
(R)-(-)-3-Aminopiperidine dihydrochloride 97%
(R)-(-)-3-Aminopiperidine dihydrochloride
Formula: C₅H₁₄Cl₂N₂
MW: 173.08 g/mol
Melting Pt: 207.5…211.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06799458
CAS Number: 334618-23-4

Order Now

Specification Test Results

Appearance White to tan Powder
Infrared spectrum Conforms
Elemental Analysis 33.5 to 35.9 % C
Elemental Analysis 15.5 to 16.8 % N
Specific optical rotation -1° to -3° (20 °C, 589 nm) (c=1, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR