(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
|
Warning
|
Synonyms:
PyAOP
, (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate
, ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)
AC44061-0050
103513-404EA
276
USD
103513-404
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Formula:
C₁₇H₂₇F₆N₇OP₂ MW: 521.4 g/mol Melting Pt: 153…155 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03703417 CAS Number: 156311-83-0 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Elemental Analysis | 38.18 to 40.14 % (C) |
Elemental Analysis | 18.33 to 19.28 % (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...