(4-Bromophenyl)(cyclopropyl)ketone 95%

Warning

Synonyms: 4-Bromophenyl Cyclopropyl ketone(4-Bromophenyl)cyclopropylmethanone(4-Bromobenzoyl)cyclopropane

AC23542-0500 AC23542-2500
103511-788EA 163 USD
103511-788 103511-790
(4-Bromophenyl)(cyclopropyl)ketone 95%
(4-Bromophenyl)(cyclopropyl)ketone
Formula: C₁₀H₉BrO
MW: 225.09 g/mol
Density: 1.45 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00019227
CAS Number: 6952-89-2

Order Now

Specification Test Results

Appearance Yellow to beige to orange Crystalline powder or low melting crystalline mass
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR