(2,3-Xylyloxy)acetic acid ≥98%

Warning

Synonyms: 2-(2,3-Dimethylphenoxy)acetic acid(2,3-Dimethylphenoxy)acetic acid

AC37247-0010 AC37247-0050
103512-942EA 40.4 USD
103512-942 103512-940
(2,3-Xylyloxy)acetic acid ≥98%
(2,3-Xylyloxy)acetic acid
Formula: (C₆H₃(CH₃)₂)OCH₂COOH
MW: 180.2 g/mol
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00016814
CAS Number: 2935-63-9

Order Now

Specification Test Results

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Melting point 186°C to 190°C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR