Miconazole nitrate

Warning

Synonyms: 1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate(±)-Miconazole nitrate salt

AC29183-0010 AC29183-0050 AC29183-0250
103512-090EA 86.7 USD
103512-090 103512-092 103512-088
Miconazole nitrate
Miconazole nitrate
Formula: C₁₈H₁₄Cl₄N₂O·HNO₃
MW: 479.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058161
CAS Number: 22832-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR