Sodium ethoxide 21% in ethanol, AcroSeal™

Supplier: ACROS ORGANICS MS

Danger

Synonyms: Sodium ethylateSodium ethanolate

AC42930-1000 AC42930-8000
103524-356EA 29.82 USD InStock
103524-356 103524-358
Sodium ethoxide 21% in ethanol, AcroSeal™
Sodium ethoxide

  • reagent for chemical synthesis

Formula: C₂H₅NaO
MW: 68,05 g/mol
Boiling Pt: 91°C -
Melting Pt: 260 °C
Density: 0.868
Flash Pt: 25°C
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Yellow to brown Solution (May darken on storage)
Appearance May darken on storage
Titration with HCl 20.0 to 22.0%
NaOH + Na2CO3 (as NaOH) =<0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR