Dimethylzinc 1.2 mol/l in toluene, AcroSeal™

Danger

AC37724-1000 AC37724-8000
103528-030EA 1255.38 USD
103528-030 103528-028
Dimethylzinc 1.2 mol/l in toluene, AcroSeal™
Dimethylzinc
Formula: C₂H₆Zn
Density: 0.93 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00014854
CAS Number: 544-97-8
EINECS: 208-884-1
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Concentration 11.5 to 12.5 wt% (On Zn basis)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR