Diethylzinc 1.5 mol/l in toluene, AcroSeal™

Danger

AC37731-8000 AC37731-1000
103528-034EA 623.08 USD
103528-034 103528-032
Diethylzinc 1.5 mol/l in toluene, AcroSeal™
Diethylzinc
Formula: C₄H₁₀Zn
Density: 0.913 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00009021
CAS Number: 557-20-0
EINECS: 209-161-3
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration Complexometric 1.4 to 1.6 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR