(4-Methoxyphenyl)magnesium bromide 1 M in THF, AcroSeal™

Danger

Synonyms: 4-Anisylmagnesium bromide

AC37742-1000
103528-048EA 62 USD
103528-048
(4-Methoxyphenyl)magnesium bromide 1 M in THF, AcroSeal™
(4-Methoxyphenyl)magnesium bromide
Formula: C₇H₇BrMgO
MW: 211.34 g/mol
Density: 1 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00013515
CAS Number: 13139-86-1
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Yellow to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.9 to 1.2 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR