(Trimethylsilyl)diazomethane ∼2.0 M in hexanes, AcroSeal™, Technical Grade

Danger

Synonyms: TrimethylsilyldiazomethaneTMS-Diazomethane

AC42921-1000
103524-350EA 568.85 USD
103524-350
(Trimethylsilyl)diazomethane ∼2.0 M in hexanes, AcroSeal™, Technical Grade
(Trimethylsilyl)diazomethane
Formula: C₄H₁₀N₂Si
MW: 114.22
Boiling Pt: 96 °C (760 mmHg)
Density: 0.675 g/cm³
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00053946
CAS Number: 18107-18-1
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow Solution
Strength 1.90 to 2.40 M
Sulfate (SO4) ≤30 ppm
Iron (Fe) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR