Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

Humidifier Filters

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (500043-146)

Supplier:  Janitorial Supplies
Description:   95% efficient HEPA-type filter
Catalog Number: (500043-145)

Supplier:  Janitorial Supplies
Description:   Helps reduce common household odors
Supplier:  Janitorial Supplies
Description:   Humidifier filter with antimicrobial treatment and air washing technology.
Catalog Number: (500043-147)

Supplier:  Janitorial Supplies
Description:   Captures 99.97% of microscopic allergens such as mold spores, pollen, dust, smoke and pet dander
Catalog Number: (102830-864)

Supplier:  Janitorial Supplies
Description:   High efficiency filter helps remove airborne particles and allergens from indoor air

Supplier:  Janitorial Supplies
Description:   Removes minerals and pollutants from water.

Supplier:  Janitorial Supplies
Description:   Replacement Carbon Filter for Harmony Air Purifiers, 4/Box
Supplier:  Janitorial Supplies
Description:   Cool mist wicking humidifier filter.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 9
  1  2  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping