Anti-Glucose 1-dehydrogenase Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91943
103279-542EA 734.58 USD
103279-542
Anti-Glucose 1-dehydrogenase Rabbit Polyclonal Antibody
Antibodies
The Glucose 1-dehydrogenase Antibody from Novus Biologicals is a rabbit polyclonal antibody to Glucose 1-dehydrogenase. This antibody reacts with human, mouse, rat. The Glucose 1-dehydrogenase Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Glucose 1-dehydrogenase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR