Anti-Gankyrin Rabbit Polyclonal Antibody

H00005716-D01P
103308-542EA 627.69 USD
103308-542
Anti-Gankyrin Rabbit Polyclonal Antibody
The Gankyrin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Gankyrin. This antibody reacts with human. The Gankyrin Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Gankyrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR