Anti-GABA Receptor Epsilon Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-21572
102712-996EA 635.51 USD
102712-996
Anti-GABA Receptor Epsilon Rabbit Polyclonal Antibody
Antibodies
The GABA Receptor Epsilon Antibody from Novus Biologicals is a rabbit polyclonal antibody to GABA Receptor Epsilon. This antibody reacts with human. The GABA Receptor Epsilon Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: GABA Receptor Epsilon
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR