Anti-Frataxin Mouse Monoclonal Antibody [clone: 3G9]

Supplier: Novus Biologicals
H00002395-M03
103326-754EA 798.46 USD
103326-754
Anti-Frataxin Mouse Monoclonal Antibody [clone: 3G9]
Antibodies
The Frataxin Antibody (3G9) from Novus Biologicals is a mouse monoclonal antibody to Frataxin. This antibody reacts with human. The Frataxin Antibody (3G9) has been validated for the following applications: Western Blot, ELISA, RNA Inhibition.

Type: Primary
Antigen: Frataxin
Clonality: Monoclonal
Clone: 3G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR