Anti-Fibrinogen gamma chain Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87448
103275-852EA 734.58 USD
103275-852
Anti-Fibrinogen gamma chain Rabbit Polyclonal Antibody
Antibodies
The Fibrinogen gamma chain Antibody from Novus Biologicals is a rabbit polyclonal antibody to Fibrinogen gamma chain. This antibody reacts with human. The Fibrinogen gamma chain Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Fibrinogen gamma chain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR