Anti-Fibrinogen gamma chain Mouse Monoclonal Antibody [clone: 1F2]

Supplier: Novus Biologicals
H00002266-M01
103326-412EA 798.46 USD
103326-412
Anti-Fibrinogen gamma chain Mouse Monoclonal Antibody [clone: 1F2]
Antibodies
The Fibrinogen gamma chain Antibody (1F2) from Novus Biologicals is a mouse monoclonal antibody to Fibrinogen gamma chain. This antibody reacts with human. The Fibrinogen gamma chain Antibody (1F2) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin, Proximity Ligation Assay.

Type: Primary
Antigen: Fibrinogen gamma chain
Clonality: Monoclonal
Clone: 1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR