Anti-Endothelin-1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00001906-D01P
103325-462EA 429 USD
103325-462
Anti-Endothelin-1 Rabbit Polyclonal Antibody
Antibodies
The Endothelin-1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Endothelin-1. This antibody reacts with human. The Endothelin-1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Endothelin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR