Anti-Dysferlin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-84695
103266-850EA 734.58 USD
103266-850
Anti-Dysferlin Rabbit Polyclonal Antibody
Antibodies
The Dysferlin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Dysferlin. This antibody reacts with human. The Dysferlin Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Dysferlin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR