Anti-EIF5 Mouse Monoclonal Antibody [clone: 1D9-4B9]

Supplier: Novus Biologicals
H00001983-M02
103325-596EA 798.46 USD
103325-596
Anti-EIF5 Mouse Monoclonal Antibody [clone: 1D9-4B9]
Antibodies
The EIF5 Antibody (1D9-4B9) from Novus Biologicals is a mouse monoclonal antibody to EIF5. This antibody reacts with human. The EIF5 Antibody (1D9-4B9) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: EIF5
Clonality: Monoclonal
Clone: 1D9-4B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR