Anti-eIF4GII Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-16309
102707-664EA 635.51 USD
102707-664
Anti-eIF4GII Rabbit Polyclonal Antibody
Antibodies
The eIF4GII Antibody from Novus Biologicals is a rabbit polyclonal antibody to eIF4GII. This antibody reacts with human, mouse, rat. The eIF4GII Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: eIF4GII
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR