Anti-eIF4GII Mouse Monoclonal Antibody [clone: 1B4]

Supplier: Novus Biologicals
H00008672-M01
103335-404EA 798.46 USD
103335-404
Anti-eIF4GII Mouse Monoclonal Antibody [clone: 1B4]
Antibodies
The eIF4GII Antibody (1B4) from Novus Biologicals is a mouse monoclonal antibody to eIF4GII. This antibody reacts with human. The eIF4GII Antibody (1B4) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: eIF4GII
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR