Anti-EAAT1/GLAST-1/SLC1A3 Goat Polyclonal Antibody

NBP2-26179
103408-466EA 466.15 USD
103408-466
Anti-EAAT1/GLAST-1/SLC1A3 Goat Polyclonal Antibody
The EAAT1 / GLAST-1 / SLC1A3 Antibody from Novus Biologicals is a goat polyclonal antibody to EAAT1 / GLAST-1 / SLC1A3. This antibody reacts with human. The EAAT1 / GLAST-1 / SLC1A3 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Type: Primary
Antigen: EAAT1/GLAST-1/SLC1A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR