Anti-DYX1C1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-20910
103408-458EA 564.74 USD
103408-458
Anti-DYX1C1 Goat Polyclonal Antibody
Antibodies
The DYX1C1 Antibody from Novus Biologicals is a goat polyclonal antibody to DYX1C1. This antibody reacts with human, canine. The DYX1C1 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Type: Primary
Antigen: DYX1C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal region
Host: Goat
Isotype: IgG
Reactivity: Human, Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR