Anti-DYRK2 Mouse Monoclonal Antibody [clone: 4G11]

Supplier: Novus Biologicals
H00008445-M03
103334-946EA 798.46 USD
103334-946
Anti-DYRK2 Mouse Monoclonal Antibody [clone: 4G11]
Antibodies
The DYRK2 Antibody (4G11) from Novus Biologicals is a mouse monoclonal antibody to DYRK2. This antibody reacts with human. The DYRK2 Antibody (4G11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: DYRK2
Clonality: Monoclonal
Clone: 4G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR