Anti-DNAJB2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-83188
103267-500EA 734.58 USD
103267-500
Anti-DNAJB2 Rabbit Polyclonal Antibody
Antibodies
The DNAJB2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DNAJB2. This antibody reacts with human. The DNAJB2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: DNAJB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR