Anti-DNAJB2 Mouse Monoclonal Antibody [clone: 1B7]

Supplier: Novus Biologicals
H00003300-M02
103328-602EA 798.46 USD
103328-602
Anti-DNAJB2 Mouse Monoclonal Antibody [clone: 1B7]
Antibodies
The DNAJB2 Antibody (1B7) from Novus Biologicals is a mouse monoclonal antibody to DNAJB2. This antibody reacts with human. The DNAJB2 Antibody (1B7) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: DNAJB2
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR