Anti-DLX5 Mouse Monoclonal Antibody [clone: 3B11]

Supplier: Novus Biologicals
H00001749-M12
103325-194EA 798.46 USD
103325-194
Anti-DLX5 Mouse Monoclonal Antibody [clone: 3B11]
Antibodies
The DLX5 Antibody (3B11) from Novus Biologicals is a mouse monoclonal antibody to DLX5. This antibody reacts with human, mouse, rat. The DLX5 Antibody (3B11) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: DLX5
Clonality: Monoclonal
Clone: 3B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR