You Searched For: Tween® 20 (Polysorbate)


16  results were found

SearchResultCount:"16"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (97063-980)
Supplier: VWR
Description: POLYSORBATE20 10%SOL PEROXID-FREE P5=1EA

Supplier: VWR
Description: Non-ionic detergent for solubilizing membrane proteins without altering biological activity.
Catalog Number: (97063-872)
Supplier: VWR
Description: Non-ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.

Supplier: MilliporeSigma
Description: Suitable for use as excipient EMPROVE® exp Ph Eur, JPE, NF
Supplier: MilliporeSigma
Description: TWEEN 20 FOR SYNTHESIS (500ML)
Supplier: TCI America
Description: CAS Number: 9005-64-5 MDL Number: MFCD00165986 Form: Clear Liquid Color: Yellow Melting point (°C): -14 Flash Point (°C): 297 Specific Gravity (20...
Supplier: TCI America
Description: [for Biochemical Research] CAS Number: 9005-64-5 MDL Number: MFCD00165986 Form: Clear Liquid Color: Yellow Melting point (°C): -14 Flash Point (°...
Supplier: BioVision
Description: A non-ionic detergent useful for the isolation of membrane-bound proteins under non-denaturing conditions.

Catalog Number: (10190-930)
Supplier: BioVision
Description: Tween* 20 Solution (10% in H2O), Alternate Name/Synonyms: Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, A 10% soultion of Tween 20 in water, 100 ml


Catalog Number: (80503-492)
Supplier: MilliporeSigma
Description: TWEEN® 20 Detergent is a molecular biology grade, pale yellow liquid that offeres a specific gravity of 1.100 to 1.110 (at 20°C)

Catalog Number: (80057-684)
Supplier: MilliporeSigma
Description: Non-ionic detergent used for immunoprecipitation and for the solubilization of membrane-bound proteins.

Catalog Number: (100216-360)
Supplier: Teknova
Description: Tween®20 is a polyoxyethylene sorbitol ester that belongs to the polysorbate family. It is a nonionic detergent having a molecular weight of 1,225 dal...


Catalog Number: (100487-942)
Supplier: Electron Microscopy Sciences
Description: TWEEN-20® is a non-ionic surfactant with hydrophobic alkyl side chains.

Minority or Woman-Owned Business Enterprise


Supplier: Promega Corporation
Description: A nonionic detergent used in many molecular biology applications.
Catalog Number: (PAW3831)
Supplier: Promega Corporation
Description: Tween® 20 is a nonionic detergent used as a buffer component for immunoscreening in the ProtoBlot Systems.

Catalog Number: (89411-094)
Supplier: Biotium
Description: Tween-20 detergent is commonly used in blocking and washing buffers for Western blotting.


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 16
no targeter for Bottom