    Sort by:
  • Search Within Results
petroleum-spirit-35-60c
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"16"
Sort by:
Supplier:  VWR International
Description:   Meets ACS specifications for general use.
SDS
Supplier:  Avantor Performance Materials
Description:   Ligroin; naphtha; petroleum naphtha. CAS RN 8032-32-4. Density: 0.63 kg/L. AR*, ACS. Actual signed Certificate of Lot Analysis provided on label. Also meets AOCS specifications. Subject to postal/shipping restrictions. 4L.
SDS
Supplier:  Avantor Performance Materials
Description:   Ligroin; naphtha; petroleum naphtha. CAS RN 8032-32-4. Density: 0.64 kg/L. ‘BAKER ANALYZED’* Reagent. Boiling range: 35-60[degree]C. Meets ACS specifications. Lot analysis on label. Packaged in a SAFETAINER*. Subject to postal/shipping restrictions. 4L.
SDS
Supplier:  Avantor Performance Materials
Description:   UltimAR™ solvents let you consolidate the number of solvent grades used in a diverse variety of applications, saving money and storage space without c...
SDS
Catalog Number: (MK498310)
Supplier:  Avantor Performance Materials
Description:   Ligroin; naphtha; petroleum naphtha. CAS RN 8032-32-4. Density: 0.63 kg/L. AR*, ACS. Actual signed Certificate of Lot Analysis provided on label. Also meets AOCS specifications. Packaged in SAFEMOR* containers. Subject to postal/shipping restrictions. 4L.
SDS
Catalog Number: (MK497104)
Supplier:  Avantor Performance Materials
Description:   Ligroin; naphtha; petroleum naphtha. CAS RN 8032-32-4. Density: 0.63 kg/L. AR*, ACS. Actual signed Certificate of Lot Analysis provided on label. Also meets AOCS specifications. Packaged in AR-CAN* containers. Subject to postal/shipping restrictions. 950mL.
SDS
Catalog Number: (MK612819)
Supplier:  Avantor Performance Materials
Description:   Ligroin; naphtha; petroleum naphtha. CAS RN 8032-32-4. For fat extraction. Boiling Range: 35-60[degree]C. Stearin Residue Test (increase in weight in g): 0.003 max. Subject to postal/shipping restrictions. 20L.
SDS
Supplier:  Avantor Performance Materials
Description:   Ligroin; naphtha; petroleum naphtha. CAS RN 8032-32-4. Density: 0.64 kg/L. ‘PHOTREX’*. Boiling Range: 35-60[degree]C. For spectrophotometry. Lot analysis on label. Packaged in a SAFETAINER. Subject to postal/shipping restrictions. 4L.
SDS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 16
  1  2  Next