    Sort by:
  • Search Within Results
Glycyrrhizic acid monoammonium salt
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   Glycyrrhizic acid monoammonium salt, 98+%, CAS number: 53956-04-0, Molecular Formula: C42 H65 N O16, Molecular weight: 839.98, Synonym: Glycamil, Color: White to light yellow, Form: Powder, Size: 10G