Anti-Calbindin D-28K Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86664
103270-286EA 734.58 USD
103270-286
Anti-Calbindin D-28K Rabbit Polyclonal Antibody
Antibodies
The Calbindin D-28K Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calbindin D-28K. This antibody reacts with human. The Calbindin D-28K Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Calbindin D-28K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR